missing translation for 'onlineSavingsMsg'
Learn More

5-Chloro-1-methyl-4-nitroimidazole, 95%

Product Code. 15468408
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15468408 5 g 5g
15458408 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15468408 Supplier Thermo Scientific Alfa Aesar Supplier No. H61824.06
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 4897-25-0 | C4H4ClN3O2 | 161.545 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4897-25-0
Molecular Formula C4H4ClN3O2
Molecular Weight (g/mol) 161.545
MDL Number MFCD00233664
InChI Key OSJUNMSWBBOTQU-UHFFFAOYSA-N
Synonym 5-chloro-1-methyl-4-nitro-1h-imidazole, 1h-imidazole, 5-chloro-1-methyl-4-nitro, 1-methyl-5-chloro-4-nitroimidazole, pcmni, imidazole, 5-chloro-1-methyl-4-nitro, unii-p917jw98bd, 1-methyl-4-nitro-5-chloroimidazole, 1-methyl-4-nitro-5-chloro imidazole, cmni, acmc-1api8
PubChem CID 21010
IUPAC Name 5-chloro-1-methyl-4-nitroimidazole
SMILES CN1C=NC(=C1Cl)[N+](=O)[O-]

Specifications

Color White
Quantity 5 g
Formula Weight 161.55
Percent Purity 95%
Physical Form Powder
Chemical Name or Material 5-Chloro-1-methyl-4-nitroimidazole
compliance-icons
Product Identifier
  • 5-Chloro-1-methyl-4-nitroimidazole
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: 5-Chloro-1-methyl-4-nitroimidazole

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.