missing translation for 'onlineSavingsMsg'
Learn More

5-Chloro-1-methyl-4-nitroimidazole, TRC

Product Code. 30132561
Change view
Click to view available options
Quantity:
500 mg
1 g
Unit Size:
1g
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30132561 500 mg 500mg
17913085 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30132561 Supplier Toronto Research Chemicals Supplier No. TRCC369270500MG
 Restricted Item
Estimated Shipment: 07-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4897-25-0
Molecular Formula C4 H4 Cl N3 O2
Molecular Weight (g/mol) 161.55
Synonym 5-Chloro-1-methyl-4-nitroimidazole
IUPAC Name 5-chloro-1-methyl-4-nitroimidazole
SMILES Cn1cnc(c1Cl)[N+](=O)[O-]

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C4H4ClN3O2/c1-7-2-6-4(3(7)5)8(9)10/h2H,1H3
Formula Weight 160.9992
Chemical Name or Material 5-Chloro-1-Methyl-4-Nitroimidazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.