missing translation for 'onlineSavingsMsg'
Learn More

5-Chloro-2(3H)-benzoxazolone, 99%, Thermo Scientific Chemicals

Product Code. 11480318
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
Unit Size:
1000g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11480318 50 g 50g
11490318 250 g 250g
11470318 1000 g 1000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11480318 Supplier Thermo Scientific Alfa Aesar Supplier No. B24507.18

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 95-25-0
Molecular Formula C7H4ClNO2
Molecular Weight (g/mol) 169.564
MDL Number MFCD00005717
InChI Key TZFWDZFKRBELIQ-UHFFFAOYSA-N
Synonym chlorzoxazone, chlorzoxazon, paraflex, chloroxazone, 5-chloro-2-benzoxazolone, biomioran, escoflex, myoflexin, myoflexine, pathorysin
PubChem CID 2733
ChEBI CHEBI:3655
IUPAC Name 5-chloro-3H-1,3-benzoxazol-2-one
SMILES C1=CC2=C(C=C1Cl)NC(=O)O2

Specifications

Melting Point 189°C to 192°C
Quantity 50 g
Merck Index 14,2194
Formula Weight 169.57
Percent Purity 99%
Chemical Name or Material 5-Chloro-2(3H)-benzoxazolone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.