missing translation for 'onlineSavingsMsg'
Learn More

5-Chloro-2-fluoroaniline, 97%

Product Code. 11470007
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11470007 50 g 50g
11460007 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11470007 Supplier Thermo Scientific Alfa Aesar Supplier No. B20951.18
 Restricted Item
Estimated Shipment: 02-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 2106-05-0 | C6H5ClFN | 145.561 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2106-05-0
Molecular Formula C6H5ClFN
Molecular Weight (g/mol) 145.561
MDL Number MFCD00069416
InChI Key JCYROOANFKVAIB-UHFFFAOYSA-N
Synonym 2-fluoro-5-chloroaniline, 5-chloro-2-fluorophenylamine, benzenamine, 5-chloro-2-fluoro, buttpark 44\01-98, 5-chlor-2-fluoranilin, pubchem1515, 5-chloro-2-fluoroani, acmc-209fh5, 5-chloro-2-fluoro-aniline, 5-chloro-2-fluorobenzenamine
PubChem CID 75015
IUPAC Name 5-chloro-2-fluoroaniline
SMILES C1=CC(=C(C=C1Cl)N)F

Specifications

Density 1.3
Quantity 50 g
Odor Characteristic
Refractive Index 1.56
UN Number UN2810
Boiling Point 102°C to 105°C (20 mmHg)
Formula Weight 145.56
Percent Purity 97%
Chemical Name or Material 5-Chloro-2-fluoroaniline

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.