missing translation for 'onlineSavingsMsg'
Learn More

5-Chloro-2-fluoropyridine-3-boronic acid, 97%, Thermo Scientific™

Catalog Number 15410088
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
937595-70-5
C5H4BClFNO2
175.35
MFCD04972408
ZUDXVHDVUTUTFH-UHFFFAOYSA-N
5-chloro-2-fluoropyridine-3-boronic acid, 5-chloro-2-fluoropyridin-3-yl boronic acid, 5-chloro-2-fluoropyridin-3-ylboronicacid, 2-fluoro-5-chloropyridine-3-boronic acid, 5-chloro-2-fluoro-3-pyridyl boronic acid, 5-chloro-2-fluoropyridine-3-boronic acid hydrate, pubchem17095, acmc-209rnf, 5-chloro-2-fluoro-3-pyridylboronic acid, 5-chloro-2-fluoro-3-pyridinyl boronic acid
24901779
(5-chloro-2-fluoropyridin-3-yl)boronic acid
B(C1=CC(=CN=C1F)Cl)(O)O
This item is not returnable. View return policy
937595-70-5
C5H4BClFNO2
175.35
MFCD04972408
ZUDXVHDVUTUTFH-UHFFFAOYSA-N
5-chloro-2-fluoropyridine-3-boronic acid, 5-chloro-2-fluoropyridin-3-yl boronic acid, 5-chloro-2-fluoropyridin-3-ylboronicacid, 2-fluoro-5-chloropyridine-3-boronic acid, 5-chloro-2-fluoro-3-pyridyl boronic acid, 5-chloro-2-fluoropyridine-3-boronic acid hydrate, pubchem17095, acmc-209rnf, 5-chloro-2-fluoro-3-pyridylboronic acid, 5-chloro-2-fluoro-3-pyridinyl boronic acid
24901779
(5-chloro-2-fluoropyridin-3-yl)boronic acid
B(C1=CC(=CN=C1F)Cl)(O)O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 125°C to 126°C
Quantity 250 mg
Formula Weight 175.35
Percent Purity 97%
Chemical Name or Material 5-Chloro-2-fluoropyridine-3-boronic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'