missing translation for 'onlineSavingsMsg'
Learn More

5-Chloro-2-nitrobenzamide, 98+%

Product Code. 11327396
Change view
Click to view available options
Quantity:
2 g
10 g
Unit Size:
10g
2g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11327396 10 g 10g
11317396 2 g 2g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11327396 Supplier Thermo Scientific Alfa Aesar Supplier No. L09380.09
 Restricted Item
Estimated Shipment: 30-07-2026
Add to basket
This item is not returnable. View return policy

CAS: 40763-96-0 | C7H5ClN2O3 | 200.578 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 40763-96-0
Molecular Formula C7H5ClN2O3
Molecular Weight (g/mol) 200.578
MDL Number MFCD00017509
InChI Key MKHXTOPPKVFSFI-UHFFFAOYSA-N
Synonym benzamide, 5-chloro-2-nitro, acmc-209jgl, 5-chloro-2-nitrobenzamid, 2-nitro-5-chlorobenzamide, 2-carbamoyl-4-chloronitrobenzene, 5-chloro-2-nitrobenzamide
PubChem CID 2723672
IUPAC Name 5-chloro-2-nitrobenzamide
SMILES C1=CC(=C(C=C1Cl)C(=O)N)[N+](=O)[O-]

Specifications

Melting Point 155°C to 159°C
Quantity 10 g
Beilstein 2267797
Formula Weight 200.58
Percent Purity ≥98%
Chemical Name or Material 5-Chloro-2-nitrobenzamide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.