missing translation for 'onlineSavingsMsg'
Learn More

5-Chloro-2-(trifluoromethoxy)benzyl bromide, 97%, Thermo Scientific™

Product Code. 15408145
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15408145 250 mg 250mg
15475367 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15408145 Supplier Thermo Scientific Alfa Aesar Supplier No. H32620.MD

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1092461-19-2
Molecular Formula C8H5BrClF3O
Molecular Weight (g/mol) 289.476
MDL Number MFCD11519358
InChI Key NVZXASWBNNGRKE-UHFFFAOYSA-N
Synonym 5-chloro-2-trifluoromethoxy benzyl bromide, 2-bromomethyl-4-chloro-1-trifluoromethoxy benzene, 5-chloro-2-trifluoromethoxy benzyl bromide, jrd
PubChem CID 57372372
IUPAC Name 2-(bromomethyl)-4-chloro-1-(trifluoromethoxy)benzene
SMILES C1=CC(=C(C=C1Cl)CBr)OC(F)(F)F

Specifications

Refractive Index 1.5
Quantity 250 mg
UN Number UN3265
Formula Weight 289.48
Percent Purity 97%
Chemical Name or Material 5-Chloro-2-(trifluoromethoxy)benzyl bromide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.