missing translation for 'onlineSavingsMsg'
Learn More

5-Chloro-3-phenyl-2,1-benzisoxazole, TRC

Product Code. 30361971
Change view
Click to view available options
Quantity:
5 g
25 g
50 g
Unit Size:
50g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30361971 25 g -
30360955 5 g 5g
30350629 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30361971 Supplier Toronto Research Chemicals Supplier No. TRCC37755525G
 Restricted Item
Estimated Shipment: 06-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 719-64-2
Molecular Formula C13H8ClNO
Molecular Weight (g/mol) 229.66
Synonym 3-Phenyl-5-chloroanthranil, 5-Chloro-3-phenyl-2,1-benzisoxazole, 5-Chloro-3-phenyl-2,1-benzisoxazoles, 5-Chloro-3-phenylanthranil, 5-Chloro-3-phenylbenz[c]isoxazole, NSC 405896
IUPAC Name 5-chloro-3-phenyl-2,1-benzoxazole
SMILES Clc1ccc2noc(c3ccccc3)c2c1

Specifications

Quantity 25 g
InChI Formula InChI=1S/C13H8ClNO/c14-10-6-7-12-11(8-10)13(16-15-12)9-4-2-1-3-5-9/h1-8H
Formula Weight 229.0294
Chemical Name or Material 5-Chloro-3-phenyl-2,1-benzisoxazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.