missing translation for 'onlineSavingsMsg'
Learn More

5-Chlorovaleronitrile, 97%, Thermo Scientific™

Product Code. 11384584
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11384584 10 g 10g
11394584 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11384584 Supplier Thermo Scientific Alfa Aesar Supplier No. L01413.09

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6280-87-1
Molecular Formula C5H8ClN
Molecular Weight (g/mol) 117.58
MDL Number MFCD00001977
InChI Key JSAWFGSXRPCFSW-UHFFFAOYSA-N
Synonym 5-chlorovaleronitrile, pentanenitrile, 5-chloro, 5-chloro-n-valeronitrile, valeronitrile, 5-chloro, 5-chloro valeronitrile, .delta.-chlorovaleronitrile, snxhfhaiuzjp@, 5-chlorovaieronitrile, 4-chlorobutyl cyanide, delta-chlorovaleronitrile
PubChem CID 80474
IUPAC Name 5-chloropentanenitrile
SMILES ClCCCCC#N

Specifications

Density 1.447
Boiling Point 92°C to 93°C (10 mmHg)
Flash Point 98°C (208°F)
Refractive Index 1.446
Quantity 10 g
UN Number UN3276
Beilstein 1742079
Formula Weight 117.58
Percent Purity 97%
Chemical Name or Material 5-Chlorovaleronitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.