missing translation for 'onlineSavingsMsg'
Learn More

5-(Difluoromethyl)-1h-pyrazol-3-amine, TRC

Product Code. 30369924
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30369924 1 g
30371475 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30369924 Supplier Toronto Research Chemicals Supplier No. TRCD4511401G
 Restricted Item
Estimated Shipment: 07-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1284220-49-0
Molecular Formula C4H5F2N3
Molecular Weight (g/mol) 133.1
SMILES Nc1cc([nH]n1)C(F)F

Specifications

Quantity 1 g
InChI Formula InChI=1S/C4H5F2N3/c5-4(6)2-1-3(7)9-8-2/h1,4H,(H3,7,8,9)
Formula Weight 133.0452
Chemical Name or Material 5-(Difluoromethyl)-1h-pyrazol-3-amine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.