Learn More
5-Ethyl-2-methylpyridine(5-Ethyl-2-picoline), TRC
Product Code. 30004029
Change view
Click to view available options
Quantity:
1 g
10 g
100 g
Unit Size:
100g
10g
1g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30004029 10 g 10g
30040709 1 g 1g
30008496 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30004029 Supplier Toronto Research Chemicals Supplier No. TRCE92553010G
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
missing translation for 'validMainframeMsg'
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 104-90-5
Molecular Formula C8 H11 N
Molecular Weight (g/mol) 121.18
Synonym Pyridine, 5-ethyl-2-methyl-, 2-Picoline, 5-ethyl- (8CI), 5-Ethyl-2-methylpyridine, 2-Methyl-5-ethylpyridine, 3-Ethyl-6-methylpyridine, 5-Ethyl-2-picoline, 5-Ethyl-α-picoline, 6-Methyl-3-ethylpyridine, Aldehydecollidine, Aldehydine, Collidine, aldehydecollidine, NSC 1984
IUPAC Name 5-ethyl-2-methylpyridine
SMILES CCc1ccc(C)nc1
Specifications
Quantity 10 g
InChI Formula InChI=1S/C8H11N/c1-3-8-5-4-7(2)9-6-8/h4-6H,3H2,1-2H3
Formula Weight 121.0891
Chemical Name or Material 5-Ethyl-2-picoline