Learn More
5-Fluoro-2,3-dihydro-1H-inden-1-one, 98%, BLD Pharm™
Product Code. 31433289
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
25 g
100 g
500 g
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
31433289 10 g
31423900 1 g
31415968 500 g
31421508 100 g
31430707 25 g
31432769 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 31433289 Supplier BLD Pharm Supplier No. BD1157110g
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Specifications
CAS 700-84-5
Molecular Formula C9H7FO
MDL Number MFCD00041031
Quantity 10 g
SMILES O=C1CCC2=C1C=CC(F)=C2
Molecular Weight (g/mol) 150.15
Percent Purity 98%
Chemical Name or Material 5-Fluoro-2,3-dihydro-1H-inden-1-one