Learn More
5-Fluoro-3-(4-piperidinyl)-1,2-benzisoxazole Hydrochloride, TRC
Product Code. 30405761
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30405761 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30405761 Supplier Toronto Research Chemicals Supplier No. TRCF595835250MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 5-Fluoro-3-(4-piperidinyl)-1,2-benzisoxazole hydrochloride
CAS 84163-16-6
Molecular Formula C12 H13 F N2 O . Cl H
Quantity 250 mg
Synonym 4-(5-Fluoro-1,2-benzisoxazol-3-yl)piperidine Hydrochloride
InChI Formula InChI=1S/C12H13FN2O.ClH/c13-9-1-2-11-10(7-9)12(15-16-11)8-3-5-14-6-4-8;/h1-2,7-8,14H,3-6H2;1H
SMILES Cl.Fc1ccc2onc(C3CCNCC3)c2c1
IUPAC Name 5-fluoro-3-piperidin-4-yl-1,2-benzoxazole;hydrochloride
Molecular Weight (g/mol) 256.7
Formula Weight 256.0779