missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Fluoro Isatin, TRC
Click to view available options
Quantity:
5 g
Unit Size:
5g
Specifications
Specifications
| Chemical Name or Material | 5-Fluoroisatin |
| CAS | 443-69-6 |
| Molecular Formula | C8 H4 F N O2 |
| Quantity | 5 g |
| Synonym | 1H-Indole-2,3-dione, 5-fluoro-, Indole-2,3-dione, 5-fluoro- (7CI,8CI), Isatin, 5-fluoro- (6CI), 5-Fluoro-1H-indole-2,3-dione, 5-Fluoro-2,3-indoledione, 5-Fluoroindoline-2,3-dione, 5-Fluoroisatin, 5-Fluoroisatine, NSC 39161 |
| InChI Formula | InChI=1S/C8H4FNO2/c9-4-1-2-6-5(3-4)7(11)8(12)10-6/h1-3H,(H,10,11,12) |
| SMILES | Fc1ccc2NC(=O)C(=O)c2c1 |
| IUPAC Name | 5-fluoro-1H-indole-2,3-dione |
| Molecular Weight (g/mol) | 165.12 |
| Formula Weight | 165.0226 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction