missing translation for 'onlineSavingsMsg'
Learn More

5-HT2C Antibody (496214) [Janelia Fluor™ 635], Novus Biologicals™

Mouse Monoclonal Antibody

Brand:  Novus Biologicals FAB5764JF635

Product Code. 30161640

  • 585.00€ / 0.10mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

5-HT2C Monoclonal antibody specifically detects 5-HT2C in Human samples. It is validated for Flow Cytometry,Immunohistochemistry,CyTOF
TRUSTED_SUSTAINABILITY
Specifications

Specifications

5-HT2C
Monoclonal
Janelia Fluor 635
5-HT1C, 5-HT-1C, 5-HT-2C, 5-HT2C5-hydroxytryptamine receptor 1C, 5-HTR2C, 5-hydroxytryptamine (serotonin) receptor 2C, 5-hydroxytryptamine receptor 2C, HTR1C, serotonin 5-HT-2C receptor, Serotonin receptor 2C
NS0 mouse myeloma cell line transfected with human 5HT2C
RUO
Primary
Human
Liquid
Flow Cytometry, Immunohistochemistry, CyTOF
496214
50mM Sodium Borate
Mouse
0.1 mL
GPCR
3358
Store at 4°C in the dark.
IgG2a
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.