missing translation for 'onlineSavingsMsg'
Learn More

5-Hydantoin acetic acid, 98%, Thermo Scientific™

Product Code. 10503781 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10503781 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10503781 Supplier Thermo Scientific Acros Supplier No. 225700050

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 5427-26-9
CAS Min % 97.5
CAS Max % 100.0
Melting Point 213°C to 216°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Molecular Formula C5H6N2O4
MDL Number MFCD00005267
Quantity 5 g
Beilstein 24, II, 136
Synonym hydantoin-5-acetic acid, 5-hydantoinacetic acid, 2,5-dioxoimidazolidin-4-yl acetic acid, 5-hydantoin acetic acid, 2-2,5-dioxoimidazolidin-4-yl acetic acid, 4-imidazolidineacetic acid, 2,5-dioxo, 2,5-dioxo-1,3-diazolidin-4-ylacetic acid, 2-2,5-dioxo-1,3-diazolidin-4-yl acetic acid, 5-carboxymethylhydantoin, acmc-20akr6
InChI Key DQQLZADYSWBCOX-UHFFFAOYNA-N
SMILES OC(=O)CC1NC(=O)NC1=O
IUPAC Name 2-(2,5-dioxoimidazolidin-4-yl)acetic acid
Molecular Weight (g/mol) 158.11
PubChem CID 95492
Formula Weight 158.11
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 5-Hydantoin acetic acid, 98%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.