missing translation for 'onlineSavingsMsg'
Learn More

5-Hydroxy-3,4-dihydronaphthalen-1(2H)-one, 98%, BLD Pharm™

Product Code. 30965041
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
10 g
25 g
100 g
Unit Size:
100g
10g
1g
250mg
25g
5g
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30965041 1 g 1g
30956994 10 g 10g
30949196 100 g 100g
30962184 25 g 25g
30961555 250 mg 250mg
30955206 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 30965041 Supplier BLD Pharm Supplier No. BD00521g
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 28315-93-7
Molecular Formula C10H10O2
MDL Number MFCD00001693
Quantity 1 g
SMILES O=C1CCCC2=C1C=CC=C2O
Molecular Weight (g/mol) 162.19
Percent Purity 98%
Chemical Name or Material 5-Hydroxy-3,4-dihydronaphthalen-1(2H)-one
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.