missing translation for 'onlineSavingsMsg'
Learn More

5-Hydroxymethyl-2'-deoxyuridine, TRC

Product Code. 30133638
Change view
Click to view available options
Quantity:
50 mg
100 mg
250 mg
Unit Size:
100mg
250mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30133638 100 mg 100mg
30138709 250 mg 250mg
17978114 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30133638 Supplier Toronto Research Chemicals Supplier No. TRCH962300100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5116-24-5
Molecular Formula C10 H14 N2 O6
Molecular Weight (g/mol) 258.228
Synonym Thymidine, α-hydroxy-, Uridine, 2'-deoxy-5-(hydroxymethyl)- (6CI,7CI,8CI), α-Hydroxythymidine, 2'-Deoxy-5-(hydroxymethyl)uridine, 2'-Desoxy-5-hydroxymethyluridine, 5-(Hydroxymethyl)-2'-desoxyuridine, 5-Hydroxymethyldeoxyuridine
IUPAC Name 1-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-5-(hydroxymethyl)pyrimidine-2,4-dione
SMILES OC[C@H]1O[C@H](C[C@@H]1O)N2C=C(CO)C(=O)NC2=O

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C10H14N2O6/c13-3-5-2-12(10(17)11-9(5)16)8-1-6(15)7(4-14)18-8/h2,6-8,13-15H,1,3-4H2,(H,11,16,17)/t6-,7+,8+/m0/s1
Formula Weight 258.0852
Chemical Name or Material 5-Hydroxymethyl-2'-deoxyuridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.