missing translation for 'onlineSavingsMsg'
Learn More

5-Hydroxytoluene-2,4-disulfonic Acid Hydrate, TRC

Product Code. 30408685
Change view
Click to view available options
Quantity:
25 mg
50 mg
250 mg
Unit Size:
250mg
25mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30408685 50 mg 50mg
30402661 250 mg 250mg
30393767 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30408685 Supplier Toronto Research Chemicals Supplier No. TRCH95451550MG
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 5-Hydroxytoluene-2,4-disulfonic Acid Hydrate
Molecular Formula C7H8O7S2•xH2O
Quantity 50 mg
Synonym m-Cresol-3,6-disulfonate Hydrate
InChI Formula InChI=1S/C7H8O7S2/c1-4-2-5(8)7(16(12,13)14)3-6(4)15(9,10)11/h2-3,8H,1H3,(H,9,10,11)(H,12,13,14)
SMILES CC1=C(S(=O)(O)=O)C=C(S(=O)(O)=O)C(O)=C1
Molecular Weight (g/mol) 268.261802
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.