missing translation for 'onlineSavingsMsg'
Learn More

5-Iodo-1,2,3-trimethoxybenzene, 95%, BLD Pharm™

Product Code. 30946514
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
250 mg
1 g
5 g
25 g
Unit Size:
100mg
1g
250mg
25g
5g
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30946514 5 g 5g
30936775 1 g 1g
30936258 100 mg 100mg
30933946 25 g 25g
30935817 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 30946514 Supplier BLD Pharm Supplier No. BD1042955g
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 25245-29-8
Molecular Formula C9H11IO3
MDL Number MFCD01318153
Quantity 5 g
SMILES COC1=C(OC)C(OC)=CC(I)=C1
Molecular Weight (g/mol) 294.09
Percent Purity 95%
Chemical Name or Material 5-Iodo-1,2,3-trimethoxybenzene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.