missing translation for 'onlineSavingsMsg'
Learn More

5-Iodouracil, 97%

Product Code. 11345249
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11345249 50 g 50g
11335249 10 g 10g
11355249 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11345249 Supplier Thermo Scientific Alfa Aesar Supplier No. A18994.18
 Restricted Item
Estimated Shipment: 27-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 696-07-1 | C4H3IN2O2 | 237.98 g/mol

It is used as an intermediate. It has an antitumor activity.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as an intermediate. It has an antitumor activity.

Solubility
Very faint turbidity in NH3aq. Soluble in 1M NaOH.

Notes
Light Sensitive. Store away from oxidizing agents and light. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 696-07-1
Molecular Formula C4H3IN2O2
Molecular Weight (g/mol) 237.98
MDL Number MFCD00006020
InChI Key KSNXJLQDQOIRIP-UHFFFAOYSA-N
Synonym 5-iodouracil, 5-iodopyrimidine-2,4 1h,3h-dione, 2,4 1h,3h-pyrimidinedione, 5-iodo, uracil, 5-iodo, 5-iodopyrimidine-2,4-diol, 2,4-dihydroxy-5-iodopyrimidine, 5-iodo uracil, unii-h59brk500m, chembl1173, 5-iodo-1,2,3,4-tetrahydropyrimidine-2,4-dione
PubChem CID 69672
ChEBI CHEBI:43636
IUPAC Name 5-iodo-1H-pyrimidine-2,4-dione
SMILES IC1=CNC(=O)NC1=O

Specifications

Melting Point 274°C to 276°C
Quantity 50 g
Beilstein 4891
Sensitivity Light sensitive
Solubility Information Very faint turbidity in NH3aq. Soluble in 1M NaOH.
Formula Weight 237.99
Percent Purity 97%
Chemical Name or Material 5-Iodouracil

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.