Learn More
Heinz Herenz 5 L Aqua Dest
Product Code. 30158515
Change view
Click to view available options
Quantity:
5 L
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30158515 5 L Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30158515 Supplier Heinz Herenz Supplier No. 1133776
  • ONLINE EXCLUSIVE PRICE
    7.55€
    4.65€ / Pack of 1
    Save 2.90€
    38% Off
View all promotions
Estimated Shipment: 01-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
5 L Aqua Dest container.
  • Single demineralized
  • For use in laboratories, beauty salons, photo studios, households and gardening.
  • Microfiltrated to be used for pharmaceutical, chemical, and technical applications
Specifications
Product Type Cleaner
Color-coded No
Form Clear Liquid
Scent Unscented
Sterility Non-sterile
Appearance Clear
Autoclavable Not Autoclavable
Color Clear
Disposable Single-use
Filtered Filtered
Flammable No
Quantity 5 L
Size 5 L Container