Learn More
5-Methyl-1H-tetrazole, 97%
Product Code. p-7058089
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
Product Code. 15406728
Brand: Thermo Scientific Alfa Aesar H60248.06
Request Bulk or Custom Format
CAS: 4076-36-2 | C2H4N4 | 84.082 g/mol
5-Methyl-1H-tetrazole is used in the preparation of novel antifungal agents based derived from N-iodopropargylazoles and N-triiodoallylazoles. 5-Methyl Tetrazole also in the preparation of novel quinoline derivatives against mycobacterium tuberculosis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
5-Methyl-1H-tetrazole is used in the preparation of novel antifungal agents based derived from N-iodopropargylazoles and N-triiodoallylazoles. 5-Methyl Tetrazole also in the preparation of novel quinoline derivatives against mycobacterium tuberculosis.
Solubility
Soluble in water.
Notes
Store at room temperature. Keep away from oxidizing agents.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 4076-36-2
Molecular Formula C2H4N4
Molecular Weight (g/mol) 84.082
MDL Number MFCD00129971
InChI Key XZGLNCKSNVGDNX-UHFFFAOYSA-N
Synonym 5-methyltetrazole, 5-methyl-1h-tetrazole, 5-methyl tetrazole, 1h-tetrazole, 5-methyl, 5-methyl-1h-tertazole, tetrazole, 5-methyl, 5-methyl-1h-1,2,3,4-tetrazole, 5-methyl-2h-1,2,3,4-tetrazole, 2h-tetrazole, 5-methyl, 5-methyl-1h-1,2,3,4-tetraazole
PubChem CID 138107
IUPAC Name 5-methyl-2H-tetrazole
SMILES CC1=NNN=N1
Specifications
Melting Point 142°C to 146°C
Quantity 5 g
Solubility Information Soluble in water.
Formula Weight 84.08
Percent Purity 97%
Chemical Name or Material 5-Methyl-1H-tetrazole
compliance-icons
Product Identifier
  • 5-Methyl-1H-tetrazole
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : 5-Methyl-1H-tetrazole