missing translation for 'onlineSavingsMsg'
Learn More

5-Methyl-2-phenyl-1,3-oxazole-4-carbaldehyde, 97%, Thermo Scientific™

Product Code. 10113653
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10113653 1 g 1g
10133284 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10113653 Supplier Thermo Scientific Maybridge Supplier No. CC26704DA
 Restricted Item
Estimated Shipment: 06-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 70170-23-9
Molecular Formula C11H9NO2
Molecular Weight (g/mol) 187.20
MDL Number MFCD08435848
InChI Key JEXONSMPSXTJFF-UHFFFAOYSA-N
Synonym 5-methyl-2-phenyloxazole-4-carbaldehyde, 4-oxazolecarboxaldehyde,5-methyl-2-phenyl, 4-oxazolecarboxaldehyde, 5-methyl-2-phenyl
PubChem CID 12471041
IUPAC Name 5-methyl-2-phenyl-1,3-oxazole-4-carbaldehyde
SMILES CC1=C(C=O)N=C(O1)C1=CC=CC=C1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 187.2
Percent Purity 97%
Chemical Name or Material 5-Methyl-2-phenyl-1, 3-oxazole-4-carbaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.