missing translation for 'onlineSavingsMsg'
Learn More

5-Methyl Chrysene (1mg/mL In Dichloromethane), TRC

Product Code. 30401322
Change view
Click to view available options
Quantity:
5 x 1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30401322 5 x 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30401322 Supplier Toronto Research Chemicals Supplier No. TRCKIT11455X1ML
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 3697-24-3
Molecular Formula C19 H14
Quantity 5 x 1 mL
Synonym Chrysene, 5-methyl-, 5-Methylchrysene, NSC 407620
InChI Formula InChI=1S/C19H14/c1-13-12-15-7-3-4-8-16(15)18-11-10-14-6-2-5-9-17(14)19(13)18/h2-12H,1H3
SMILES Cc1cc2ccccc2c3ccc4ccccc4c13
IUPAC Name 5-methylchrysene
Molecular Weight (g/mol) 242.31
Formula Weight 242.1096
Concentration or Composition (by Analyte or Components) 5-Methylchrysene 1 mg/mL
Chemical Name or Material 5-Methylchrysene
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.