missing translation for 'onlineSavingsMsg'
Learn More

5-Methylisoxazole-4-carboxylic acid, 98+%, Thermo Scientific™

Product Code. 11313626
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11313626 25 g 25g
11303626 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11313626 Supplier Thermo Scientific Alfa Aesar Supplier No. L14628.14

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 42831-50-5
Molecular Formula C5H4NO3
Molecular Weight (g/mol) 126.09
MDL Number MFCD00955655
InChI Key VQBXUKGMJCPBMF-UHFFFAOYSA-M
Synonym 5-methylisoxazole-4-carboxylic acid, 5-methyl-4-isoxazolecarboxylic acid, unii-bl2q213q2m, 4-isoxazolecarboxylic acid, 5-methyl, 5-methylisoxazole-4-carboxylicacid, leflunomide impurity d ep, pubchem8652, leflunomide impurity d, acmc-209jr8, ksc235o3b
PubChem CID 1425240
SMILES CC1=C(C=NO1)C([O-])=O

Specifications

Melting Point 144°C to 149°C
Quantity 25 g
Beilstein 116000
IUPAC Name 5-methyl-1,2-oxazole-4-carboxylic acid
Formula Weight 127.1
Percent Purity ≥98%
Chemical Name or Material 5-Methylisoxazole-4-carboxylic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.