missing translation for 'onlineSavingsMsg'
Learn More

5-Methylisoxazole-4-carboxylic Acid, TRC

Product Code. 30474841
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30474841 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30474841 Supplier Toronto Research Chemicals Supplier No. TRCM3150001G
 Restricted Item
Estimated Shipment: 07-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 42831-50-5
Molecular Formula C5 H5 N O3
Quantity 1 g
Synonym 5-Methylisoxazole-4-carboxylic Acid, Leflunomide Imp. D (EP)
InChI Formula InChI=1S/C5H5NO3/c1-3-4(5(7)8)2-6-9-3/h2H,1H3,(H,7,8)
SMILES Cc1oncc1C(=O)O
IUPAC Name 5-methyl-1,2-oxazole-4-carboxylic acid
Molecular Weight (g/mol) 127.1
Formula Weight 127.0269
Chemical Name or Material 5-Methylisoxazole-4-carboxylic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.