missing translation for 'onlineSavingsMsg'
Learn More

5-Methylpyridine-2-boronic acid, 95%

Product Code. 15459257
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15459257 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15459257 Supplier Thermo Scientific Alfa Aesar Supplier No. H52517.MD
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 372963-49-0 | C6H8BNO2 | 136.945 g/mol

5-Methylpyridine-2-boronic acid is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Methylpyridine-2-boronic acid is used as an organic chemical synthesis intermediate.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxid
TRUSTED_SUSTAINABILITY

Specifications

CAS 372963-49-0
Melting Point 86°C to 90°C
Molecular Formula C6H8BNO2
MDL Number MFCD00674179
Quantity 250 mg
Synonym 5-methylpyridine-2-boronic acid, 5-methylpyridin-2-yl boronic acid, 5-methyl-2-pyridineboronic acid, 5-picoline-2-boronic acid, boronic acid, 5-methyl-2-pyridinyl, 5-methyl-2-pyridinyl-boronic acid, pubchem6938, acmc-209is4, 5-methyl-2-pyridyl boronic acid, 5-methyl-2-pyridinyl boronic acid
Solubility Information Slightly soluble in water.
InChI Key VDRIXWWGBDNWCM-UHFFFAOYSA-N
SMILES B(C1=NC=C(C=C1)C)(O)O
IUPAC Name (5-methylpyridin-2-yl)boronic acid
Molecular Weight (g/mol) 136.945
PubChem CID 2762743
Formula Weight 136.95
Percent Purity 95%
Chemical Name or Material 5-Methylpyridine-2-boronic acid
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.