Learn More
5-Methyltryptamine Hydrochloride, TRC
Product Code. 30480850
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30480850 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30480850 Supplier Toronto Research Chemicals Supplier No. TRCM3319401G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 5-Methyltryptamine Hydrochloride
CAS 1010-95-3
Molecular Formula C11H15ClN2
Quantity 1 g
Synonym 5-Methyl-1H-Indole-3-ethanamine Hydrochloride, 3-(2-Aminoethyl)-5-methylindole Hydrochloride
InChI Formula InChI=1S/C11H14N2.ClH/c1-8-2-3-11-10(6-8)9(4-5-12)7-13-11;/h2-3,6-7,13H,4-5,12H2,1H3;1H
SMILES Cl.Cc1ccc2[nH]cc(CCN)c2c1
IUPAC Name 2-(5-methyl-1H-indol-3-yl)ethanamine;hydrochloride
Molecular Weight (g/mol) 210.7
Formula Weight 210.0924