missing translation for 'onlineSavingsMsg'
Learn More

5-Nonanol, 98%

Product Code. 15408849
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15408849 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15408849 Supplier Thermo Scientific Alfa Aesar Supplier No. L05812.14
 Restricted Item
Estimated Shipment: 02-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 623-93-8 | C9H20O | 144.258 g/mol

5-Nonanol is used as intermediates in chemical research. It is used as natural substances and extractives.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Nonanol is used as intermediates in chemical research. It is used as natural substances and extractives.

Solubility
Soluble in water (0.46 mg/ml at 15°C), and chloroform.

Notes
Incompatible ma
TRUSTED_SUSTAINABILITY

Specifications

CAS 623-93-8
Melting Point 5°C
Density 0.826
Boiling Point 194°C to 196°C
Flash Point 96°C (204°F)
Molecular Formula C9H20O
Refractive Index 1.429
MDL Number MFCD00021944
Quantity 25 g
Beilstein 1733552
Synonym 5-nonanol, dibutylcarbinol, dibutyl carbinol, 5-nonyl alcohol, nonanol-5, acmc-1bc2t, di-n-butyl carbinol, 5-nonanol 25g, 5-nonanol gc, 5-nonanol, puriss gc
Solubility Information Soluble in water (0.46mg/ml at 15°C),and chloroform.
InChI Key FCBBRODPXVPZAH-UHFFFAOYSA-N
SMILES CCCCC(CCCC)O
IUPAC Name nonan-5-ol
Molecular Weight (g/mol) 144.258
PubChem CID 12202
Formula Weight 144.26
Percent Purity 98%
Chemical Name or Material 5-Nonanol
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.