missing translation for 'onlineSavingsMsg'
Learn More

5-Norbornene-2-carboxylic acid, 98%, mixture of endo and exo

Product Code. 15299876 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15299876 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15299876 Supplier Thermo Scientific Acros Supplier No. 453300250
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 120-74-1 | C8H10O2 | 138.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 120-74-1
Color Colorless to Yellow
Density 1.1290g/mL
Boiling Point 136.0°C to 138.0°C (14.0 mmHg)
Flash Point >110°C
Assay Percent Range 97.5% min. (total of isomers) (GC)
Odor Stench
Molecular Formula C8H10O2
Refractive Index 1.4930 to 1.4960 (20°C, 589 nm)
Quantity 25 g
UN Number 3265
Specific Gravity 1.129
Synonym 5-norbornene-2-carboxylic acid, bicyclo 2.2.1 hept-5-ene-2-carboxylic acid, norbornenecarboxylic acid, exo-5-norbornene-2-carboxylic acid, 5-norbornene-2-carboxylic acid, exo, 5-norbornene-2-carboxylic acid 8ci, bicyclo 2.2.1 hept-2-ene-5-carboxylic acid, bicyclo 2.2.1-5-heptene-2-carboxylic acid, norborn-5-ene-2-carboxylic acid
InChI Key FYGUSUBEMUKACF-UHFFFAOYSA-N
SMILES C1C2CC(C1C=C2)C(=O)O
IUPAC Name bicyclo[2.2.1]hept-2-ene-5-carboxylic acid
Molecular Weight (g/mol) 138.16
PubChem CID 78949
Formula Weight 138.16
Percent Purity 98%
Concentration or Composition (by Analyte or Components) Bicyclo[2.2.1]hept-5-ene-2-carboxylic acid
Physical Form Liquid or Solid
Chemical Name or Material 5-Norbornene-2-Carboxylic Acid
Show More Show Less
compliance-icons
Product Identifier
  • 5-Norbornene-2-carboxylic acid
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.