missing translation for 'onlineSavingsMsg'
Learn More

5-O-Desmethyl Omeprazole, TRC

Product Code. 30011586
Change view
Click to view available options
Quantity:
2.5 mg
5 mg
50 mg
Unit Size:
2.5mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30011586 5 mg 5mg
30009908 2.5 mg 2.5mg
30012798 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30011586 Supplier Toronto Research Chemicals Supplier No. TRCD2921205MG
 Restricted Item
Estimated Shipment: 04-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 151602-49-2
Molecular Formula C16H17N3O3S
Molecular Weight (g/mol) 331.39
Synonym 1H-Benzimidazol-6-ol, 2-[[(4-methoxy-3,5-dimethyl-2-pyridinyl)methyl]sulfinyl]-, 2-[(4-methoxy-3,5-dimethyl-2-pyridyl)methylsulfinyl]-1H-benzimidazol-5-ol
SMILES COc1c(C)cnc(CS(=O)c2nc3cc(O)ccc3[nH]2)c1C

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C16H17N3O3S/c1-9-7-17-14(10(2)15(9)22-3)8-23(21)16-18-12-5-4-11(20)6-13(12)19-16/h4-7,20H,8H2,1-3H3,(H,18,19)
Formula Weight 331.0991
Percent Purity >95%
Chemical Name or Material 5-O-Desmethyl Omeprazole
Grade HPLC
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.