missing translation for 'onlineSavingsMsg'
Learn More

5-Oxo Atorvastatin, TRC

Product Code. 30008027
Change view
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30008027 1 mg 1mg
30007918 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30008027 Supplier Toronto Research Chemicals Supplier No. TRCO8471551MG
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1391052-82-6
Molecular Formula C33 H33 F N2 O5
Molecular Weight (g/mol) 556.62
Synonym (βR)-2-(4-Fluorophenyl)-β-hydroxy-5-(1-methylethyl)-δ-oxo-3-phenyl-4-[(phenylamino)carbonyl]-1H-pyrrole-1-heptanoic Acid, (3R)-7-[2-(4-Fluorophenyl)-3-phenyl-4-(phenylcarbamoyl)-5-propan-2-ylpyrrol-1-yl]-3-hydroxy-5-oxoheptanoic Acid
IUPAC Name (3R)-7-[2-(4-fluorophenyl)-3-phenyl-4-(phenylcarbamoyl)-5-propan-2-ylpyrrol-1-yl]-3-hydroxy-5-oxoheptanoic acid
SMILES CC(C)c1c(C(=O)Nc2ccccc2)c(c3ccccc3)c(c4ccc(F)cc4)n1CCC(=O)C[C@@H](O)CC(=O)O

Specifications

Quantity 1 mg
InChI Formula InChI=1S/C33H33FN2O5/c1-21(2)31-30(33(41)35-25-11-7-4-8-12-25)29(22-9-5-3-6-10-22)32(23-13-15-24(34)16-14-23)36(31)18-17-26(37)19-27(38)20-28(39)40/h3-16,21,27,38H,17-20H2,1-2H3,(H,35,41)(H,39,40)/t27-/m1/s1
Formula Weight 556.2374
Chemical Name or Material 5-Oxo Atorvastatin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.