missing translation for 'onlineSavingsMsg'
Learn More

5-Oxorosuvastatin Methyl Ester, Mikromol™

Product Code. 30050074
Change view
Click to view available options
Quantity:
25 mg
100 mg
4 x 25 mg
Unit Size:
100mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30050074 25 mg 25mg
17960496 100 mg 100mg
30459233 4 x 25 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30050074 Supplier Mikromol Supplier No. MM0936.030025

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

5-Oxorosuvastatin Methyl Ester
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 5-Oxorosuvastatin Methyl Ester
CAS 147118-39-6
Analyte or Component Names 5-Oxorosuvastatin Methyl Ester
Shipping Condition Room Temperature
Molecular Formula C23 H28 F N3 O6 S
Certifications/Compliance ISO 17025
Synonym 5-Oxorosuvastatin Methyl Ester, 6-Heptenoic acid, 7-[4-(4-fluorophenyl)-6-(1-methylethyl)-2-[methyl(methylsulfonyl)amino]-5-pyrimidinyl]-3-hydroxy-5-oxo-, methyl ester, (3R,6E)-, 6-Heptenoic acid, 7-[4-(4-fluorophenyl)-6-(1-methylethyl)-2-[methyl(methylsulfonyl)amino]-5-pyrimidinyl]-3-hydroxy-5-oxo-, methyl ester, [R-(E)]-, Methyl (+)-(3R)-7-[4-(4-fluorophenyl)-6-isopropyl-2-(N-methyl-N-Methansulphonylamino)pyrimidin-5-yl]-3-hydroxy-5-oxo-6(E)-heptenoate, Methyl (3R,6E)-7-{4-(4-fluorophenyl)-6-isopropyl-2-[methyl(methylsulfonyl)amino]-5-pyrimidinyl}-3-hydroxy-5-oxo-6-heptenoate
InChI Formula InChI=1S/C23H28FN3O6S/c1-14(2)21-19(11-10-17(28)12-18(29)13-20(30)33-4)22(15-6-8-16(24)9-7-15)26-23(25-21)27(3)34(5,31)32/h6-11,14,18,29H,12-13H2,1-5H3/b11-10+/t18-/m1/s1
SMILES COC(=O)C[C@H](O)CC(=O)\C=C\c1c(nc(nc1c2ccc(F)cc2)N(C)S(=O)(=O)C)C(C)C
IUPAC Name methyl (E,3R)-7-[4-(4-fluorophenyl)-2-[methyl(methylsulfonyl)amino]-6-propan-2-ylpyrimidin-5-yl]-3-hydroxy-5-oxohept-6-enoate
Molecular Weight (g/mol) 493.55
Quantity 25 mg
Formula Weight 493.1683 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.