missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 5-tert-Butyl-2-methoxyaniline, 97%

Product Code. 15401138
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15401138 5 g 5g
15411138 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15401138 Supplier Alfa Aesar™ Supplier No. H55019.06
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 3535-88-4
Molecular Formula C11H17NO
Molecular Weight (g/mol) 179.263
MDL Number MFCD00026299
InChI Key DLTLFDQLMFMTRQ-UHFFFAOYSA-N
Synonym 5-tert-butyl-2-methoxyaniline, benzenamine, 5-1,1-dimethylethyl-2-methoxy, 4-tert-butyl-o-anisidine, 2-amine-4-tert-butylanisole, 5-tert-butyl-o-anisidine, 5-tert-butyl-2-methoxy-phenylamine, 5-tert-butyl-2-methoxyphenylamine
PubChem CID 4176318
IUPAC Name 5-tert-butyl-2-methoxyaniline
SMILES CC(C)(C)C1=CC(=C(C=C1)OC)N

Specifications

Melting Point 29°C to 30°C
Density 0.988
Boiling Point 204°C to 208°C
Flash Point 113°C (235°F)
Quantity 5 g
Formula Weight 179.26
Percent Purity 97%
Chemical Name or Material 5-tert-Butyl-2-methoxyaniline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.