Learn More
Hamilton™ 500 μL, Model 1750 C SYR, 1/4-28 Threads
Click to view available options
Volume (Metric):
500 μL
Unit Size:
Each
Description
Specification Sheet
- Needle Sold Separately – Yes
- Volume – 500 μL
- Syringe Type – Gastight
- Syringe Series – 1700
- Termination – ChemSeal (C, 1/4 in.-28)
- Needle – No Needle Available
- Fluid Path – Borosilicate glass, PTFE
- Autoclavable – Yes
- Gas Sterilizable – Yes
- Pressure – 700 psig (48.3 bar)
- Maximum Temperature – 50°C / 122°F
- Minimum Temperature – 20°C / 68°F
- Sales Quantity – 1 syringe per pack
Specifications
Specifications
| Product Type | Gastight |
| Volume (Metric) | 500 μL |