missing translation for 'onlineSavingsMsg'
Learn More

Glycerol Triformate, TRC

Product Code. 30010775
Change view
Click to view available options
Quantity:
500 mg
5 g
Unit Size:
500mg
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30010775 500 mg 500mg
30135384 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30010775 Supplier Toronto Research Chemicals Supplier No. TRCG601545500MG
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 32765-69-8
Molecular Formula C6H8O6
Molecular Weight (g/mol) 176.12
Synonym Triformin, 1,2,3-Propanetriol, 1,2,3-Triformate, 1,2,3-Propanetriol, Triformate
IUPAC Name 2,3-diformyloxypropyl formate
SMILES O=COCC(COC=O)OC=O

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C6H8O6/c7-3-10-1-6(12-5-9)2-11-4-8/h3-6H,1-2H2
Formula Weight 176.0321
Percent Purity >95%
Chemical Name or Material Glycerol Triformate
Grade GC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.