missing translation for 'onlineSavingsMsg'
Learn More

2-(((5-((Dimethylamino)methyl)furan-2-yl)methyl)thio)ethanamine Fumarate (2:1), TRC

Product Code. 30137157
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30137157 50 mg 50mg
30011692 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30137157 Supplier Toronto Research Chemicals Supplier No. TRCD46173550MG
 Restricted Item
Estimated Shipment: 18-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 256948-32-0
Molecular Formula 2 C10 H18 N2 O S . C4 H4 O4
Molecular Weight (g/mol) 544.7276
Synonym 5-[[(2-aminoethyl)thio]methyl]-N,N-dimethyl-2-furanmethanamine (2E)-2-butenedioate (2:1), USP Ranitidine Related Compound A
IUPAC Name (E)-but-2-enedioic acid;2-[[5-[(dimethylamino)methyl]furan-2-yl]methylsulfanyl]ethanamine
SMILES CN(C)Cc1oc(CSCCN)cc1.CN(C)Cc2oc(CSCCN)cc2.OC(=O)\C=C\C(=O)O

Specifications

Quantity 50 mg
InChI Formula InChI=1S/2C10H18N2OS.C4H4O4/c2*1-12(2)7-9-3-4-10(13-9)8-14-6-5-11;5-3(6)1-2-4(7)8/h2*3-4H,5-8,11H2,1-2H3;1-2H,(H,5,6)(H,7,8)/b;;2-1+
Formula Weight 544.2389
Chemical Name or Material 2-[([5-[(Dimethylamino)methyl]furan-2-yl]methyl)sulfanyl]ethanamine hemifumarate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.