missing translation for 'onlineSavingsMsg'
Learn More

N-(4-Chlorobenzyl)-1-(5-nitrothiophen-2-yl)methanamine, TRC

Product Code. 30008347
Change view
Click to view available options
Quantity:
50 mg
500 mg
Unit Size:
500mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30008347 50 mg 50mg
30135854 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30008347 Supplier Toronto Research Chemicals Supplier No. TRCC36423550MG
 Restricted Item
Estimated Shipment: 21-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1384516-10-2
Molecular Formula C12H11ClN2O2S
Molecular Weight (g/mol) 282.75
Synonym N-[(4-Chlorophenyl)methyl]-5-nitro-2-thiophenemethanamine (ACI), N-(4-Chlorobenzyl)-1-(5-nitrothiophen-2-yl)methanamine
IUPAC Name N-(4-chlorobenzyl)-1-(5-nitrothiophen-2-yl)methanamine
SMILES O=[N+](C1=CC=C(CNCC2=CC=C(Cl)C=C2)S1)[O-]

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C12H11ClN2O2S/c13-10-3-1-9(2-4-10)7-14-8-11-5-6-12(18-11)15(16)17/h1-6,14H,7-8H2
Formula Weight 282.02
Percent Purity >95%
Chemical Name or Material N-(4-Chlorobenzyl)-1-(5-nitrothiophen-2-yl)methanamine
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.