missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bromoethane-2,2,2-d3, CDN
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Specifications
Specifications
| Chemical Name or Material | Bromoethane-d3 |
| CAS | 7439-86-3 |
| Molecular Formula | C2 D3 H2 Br |
| Quantity | 5 g |
| Synonym | Ethyl Bromide-d3, 2-Bromoethane-2,2,2-d3;Bromoethane-d3, Ethyl-d3 Bromide, Trideuterioethyl Bromide-d3, 2,2,2-Trideuteroethyl Bromide, 2-Bromo-1,1,1-trideuteroethane |
| InChI Formula | InChI=1 S/C2H5Br/c1-2-3/h2H2,1H3/I1d3 |
| SMILES | [2 H]C([2 H])([2 H])CBr |
| IUPAC Name | 2-bromo-1,1,1-trideuterioethane |
| Molecular Weight (g/mol) | 111.98 |
| Formula Weight | 110.9763 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction