Learn More
iso-Propyl-d7 Alcohol, CDN
Click to view available options
Quantity:
5 g
Unit Size:
5g
Specifications
Specifications
| Chemical Name or Material | Isopropanol-d7 |
| CAS | 19214-96-1 |
| Molecular Formula | C3 2H7 H O |
| Quantity | 5 g |
| InChI Formula | InChI=1 S/C3H8O/c1-3(2)4/h3-4 H,1-2H3/i1D3,2D3,3 D |
| SMILES | [2 H]C([2 H])([2 H])C([2 H])(O)C([2 H])([2 H])[2 H] |
| IUPAC Name | 1,1,1,2,3,3,3-heptadeuteriopropan-2-ol |
| Molecular Weight (g/mol) | 67.14 |
| Formula Weight | 67.1015 g/mol |
| Percent Purity | 99 atom % D, min 98% Chemical Purity |