missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ 5me-dC Phosphoramidite, standard grade, serum vial bottle

Product Code. 18054090 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18054090 1 g 1g
17977172 5 g Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18054090 Supplier Thermo Scientific™ Supplier No. 27174804
 Restricted Item
In Stock
Add to basket
Add to basket
This item is not returnable. View return policy

5me-dC CEP is a phosphoramidite containing 5' DMT and the nucleoside deoxycytidine (dC) with Benzoyl (Bz) base protection, for use in oligonucleotide synthesis.

5me-dC CEP is a phosphoramidite containing 5' DMT and the nucleoside deoxycytidine (dC) with Benzoyl (Bz) base protection, for use in oligonucleotide synthesis. The 5-me-dC Phosphoramidite elicits enhanced binding and increased duplex stabilization (to dG) compared to unmethylated dC. This can be attributed to the hydrophobic nature of the methyl groups that help eliminate water molecules from the duplex. Thus, it can be utilized to improve the ability of an oligo to hybridize to its target. It has proven especially useful in oligo applications such as strong-binding PCR primers, immune stimulation (via antisense), and in methylation studies. This product is supplied in a 20 mL Amber bottle with a Serum Vial neck style, which is compatible with most ABI Systems|AKTA Systems of oligonucleotide synthesizers.

Features:
Full chemical name: 5'-O-(4,4'-Dimethoxytrityl)-N4-Benzoyl-5-Methyl-2'-deoxycytidine-3'-O-[(2-cyanoethyl)-(N,N-diisopropyl)]-phosphoramidite
Molecular formula: C47H54N5O8P
Base: 5me-dC
Base Protection: Bz
Formula weight: 847.95 g/mol
CAS number: 105931-57-5
HPLC Purity: >98%; M-11 Impurity ≤0.01%
31P NMR Purity: ≥98%; HMT impurity (∼146ppm): ≤0.10%

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Equipment) AKTA Synthesizers, Applied Biosystems Synthesizers
Product Type 5me-dC CEP
Content And Storage Store in freezer (at or below -20°C).
Nucleoside Type Deoxycytidine (dC)
Shipping Condition Room Temperature
Purity 98%
Quantity 1 g
Reagent Type Phosphoramidite
Grade Standard
Nucleic Acid Type DNA
Form Powder
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.