missing translation for 'onlineSavingsMsg'
Learn More

(5R)-[3-(3-Fluoro-4-morpholinophenyl)-2-oxooxazolidin-5-yl]methyl Methanesulfonate, Mikromol™

Product Code. 30050731
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30050731 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30050731 Supplier Mikromol Supplier No. MM3300.060025
 Restricted Item
Estimated Shipment: 28-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

(5R)-[3-(3-Fluoro-4-morpholinophenyl)-2-oxooxazolidin-5-yl]methyl Methanesulfonate
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (5R)-[3-(3-Fluoro-4-morpholinophenyl)-2-oxooxazolidin-5-yl]methyl Methanesulfonate
CAS 174649-09-3
Analyte or Component Names (5R)-[3-(3-Fluoro-4-morpholinophenyl)-2-oxooxazolidin-5-yl]methyl Methanesulfonate
Shipping Condition Room Temperature
Molecular Formula C15 H19 F N2 O6 S
Certifications/Compliance ISO 17025
Synonym (R)-[3-(3-Fluoro-4-morpholinophenyl)-2-oxooxazolidin-5-yl]methyl methanesulfonate, (5R)-3-[3-Fluoro-4-(4-morpholinyl)phenyl]-5-[[(methylsulfonyl)oxy]methyl]-2-oxazolidinone, 2-Oxazolidinone, 3-[3-fluoro-4-(4-morpholinyl)phenyl]-5-[[(methylsulfonyl)oxy]methyl]-, (5R)-, 2-Oxazolidinone, 3-[3-fluoro-4-(4-morpholinyl)phenyl]-5-[[(methylsulfonyl)oxy]methyl]-, (R)-, (R)-[3-(3-Fluoro-4-morpholin-4-ylphenyl)-2-oxo-5-oxazolidinyl]methyl methanesulfonate, Linezolid Related Compound D, Linezolid Related Compound D
InChI Formula InChI=1S/C15H19FN2O6S/c1-25(20,21)23-10-12-9-18(15(19)24-12)11-2-3-14(13(16)8-11)17-4-6-22-7-5-17/h2-3,8,12H,4-7,9-10H2,1H3/t12-/m1/s1
SMILES CS(=O)(=O)OC[C@H]1CN(C(=O)O1)c2ccc(N3CCOCC3)c(F)c2
IUPAC Name [(5R)-3-(3-fluoro-4-morpholino-phenyl)-2-oxo-oxazolidin-5-yl]methyl methanesulfonate
Molecular Weight (g/mol) 374.38
Quantity 25 mg
Formula Weight 374.0948 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.