missing translation for 'onlineSavingsMsg'
Learn More

(5R-cis)-Toluene-4-sulfonic Acid 5-(2,4-Difluorophenyl)-5-[1,2,4]triazol-1-ylmethyltetrahydrofuran-3-ylmethyl Ester, TRC

Product Code. 30457317
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30457317 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30457317 Supplier Toronto Research Chemicals Supplier No. TRCT53590050MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (5R-cis)-Toluene-4-sulfonic Acid 5-(2,4-Difluorophenyl)-5-[1,2,4]triazol-1-ylmethyltetrahydrofuran-3-ylmethyl Ester
CAS 149809-43-8
Molecular Formula C21 H21 F2 N3 O4 S
Quantity 50 mg
Synonym 2,5-Anhydro-1,3,4-trideoxy-2-C-(2,4-difluorophenyl)-4-[[[(4-methylphenyl)sulfonyl]oxy]methyl]-1-(1H-1,2,4-triazol-1-yl)-D-threo-pentitol, Posaconazole Impurity
InChI Formula InChI=1S/C21H21F2N3O4S/c1-15-2-5-18(6-3-15)31(27,28)30-11-16-9-21(29-10-16,12-26-14-24-13-25-26)19-7-4-17(22)8-20(19)23/h2-8,13-14,16H,9-12H2,1H3/t16-,21-/m0/s1
SMILES Cc1ccc(cc1)S(=O)(=O)OC[C@@H]2CO[C@](Cn3cncn3)(C2)c4ccc(F)cc4F
IUPAC Name [(3S,5R)-5-(2,4-difluorophenyl)-5-(1,2,4-triazol-1-ylmethyl)oxolan-3-yl]methyl 4-methylbenzenesulfonate
Molecular Weight (g/mol) 449.47
Formula Weight 449.1221
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.