missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ 5T4 Recombinant Rabbit Monoclonal Antibody (24GB2035)

Rabbit Recombinant Monoclonal Antibody

Brand:  Invitrogen™ MA557805

 View more versions of this product

Product Code. 30276155

  • 198.00€ / 20µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

5T4 Recombinant Monoclonal Antibody for Western Blot, Flow

This gene encodes a leucine-rich transmembrane glycoprotein that may be involved in cell adhesion. The encoded protein is an oncofetal antigen that is specific to trophoblast cells. In adults this protein is highly expressed in many tumor cells and is associated with poor clinical outcome in numerous cancers. Alternate splicing in the 5' UTR results in multiple transcript variants that encode the same protein. [provided by RefSeq, Oct 2009].
TRUSTED_SUSTAINABILITY
Specifications

Specifications

5T4
Recombinant Monoclonal
Unconjugated
TPBG
5T4; 5T4 oncofetal antigen; 5T4 oncofetal trophoblast glycoprotein; 5T4 oncotrophoblast glycoprotein; 5T4AG; 5T4-AG; AW495680; M6P1; TPBG; Trophoblast glycoprotein; WAIF1; Wnt-activated inhibitory factor 1
Rabbit
Affinity Chromatography
RUO
7162
-20°C
Liquid
Flow Cytometry, Western Blot
24GB2035
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Q13641
TPBG
A synthesized peptide derived from human 5T4 (aa 110-170).
20 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.