Learn More
(5Z,8Z,11Z,14Z)-Icosa-5,8,11,14-tetraenoic acid, 99%, BLD Pharm™
Product Code. 31004177
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
250 mg
1 g
5 g
10 g
Unit Size:
100mg
10g
1g
250mg
5g
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
31004177 100 mg 100mg
30997994 1 g 1g
30989106 250 mg 250mg
31001058 10 g 10g
30999956 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 31004177 Supplier BLD Pharm Supplier No. BD114601100mg
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Specifications
CAS 506-32-1
Molecular Formula C20H32O2
MDL Number MFCD00004417
Quantity 100 mg
SMILES CCCCC/C=C\C/C=C\C/C=C\C/C=C\CCCC(O)=O
Molecular Weight (g/mol) 304.47
Percent Purity 99%
Chemical Name or Material (5Z,8Z,11Z,14Z)-Icosa-5,8,11,14-tetraenoic acid