missing translation for 'onlineSavingsMsg'
Learn More

6-(4-Methoxyphenyl)-2-naphthoic Acid, TRC

Product Code. 30483024
Change view
Click to view available options
Quantity:
100 mg
1 g
Unit Size:
100mg
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30483024 100 mg 100mg
30469710 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30483024 Supplier Toronto Research Chemicals Supplier No. TRCM266125100MG
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 132292-17-2
Molecular Formula C18H14O3
Molecular Weight (g/mol) 278.3
Synonym 2-(4-Methoxyphenyl)naphthalene-6-carboxylic Acid, 6-(4-Methoxyphenyl)-2-naphthalenecarboxylic Acid
IUPAC Name 6-(4-methoxyphenyl)naphthalene-2-carboxylic acid
SMILES COc1ccc(cc1)c2ccc3cc(ccc3c2)C(=O)O

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C18H14O3/c1-21-17-8-6-12(7-9-17)13-2-3-15-11-16(18(19)20)5-4-14(15)10-13/h2-11H,1H3,(H,19,20)
Formula Weight 278.0943
Chemical Name or Material 6-(4-Methoxyphenyl)-2-naphthoic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.