Learn More
6,6'-Dibromo[1,1'-binaphthalene]-2,2'-diol, 98%, BLD Pharm™
Product Code. 31091896
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
25 g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
31091896 5 g
31078348 250 mg
31089060 25 g
31087766 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 31091896 Supplier BLD Pharm Supplier No. BD11205g
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Specifications
CAS 13185-00-7
Molecular Formula C20H12Br2O2
MDL Number MFCD00798290
Quantity 5 g
SMILES OC1=CC=C2C=C(Br)C=CC2=C1C3=C4C=CC(Br)=CC4=CC=C3O
Molecular Weight (g/mol) 444.12
Percent Purity 98%
Chemical Name or Material 6,6'-Dibromo[1,1'-binaphthalene]-2,2'-diol