missing translation for 'onlineSavingsMsg'
Learn More

(6,7-Dihydro-4H-thieno[3,2-c]pyran-4-yl)methylamine, 97%, Thermo Scientific™

Product Code. p-3484505
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10133824 250 mg Amber Glass Bottle 250mg
10193864 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10133824 Supplier Thermo Scientific Maybridge Supplier No. CC61114CB

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 63932-26-3
Molecular Formula C8H11NOS
Molecular Weight (g/mol) 169.242
MDL Number MFCD08690319
InChI Key NMVKUIFBCHQTHO-UHFFFAOYSA-N
Synonym 4h,6h,7h-thieno 3,2-c pyran-4-ylmethanamine, 6,7-dihydro-4h-thieno 3,2-c pyran-4-yl methylamine, 4h-thieno 3,2-c pyran-4-methanamine,6,7-dihydro, 6,7-dihydro-4h-thieno-3,2-c pyran-4-ylmethyl amine, 6,7-dihydro-4h-thieno 3,2-c pyran-4-yl methanamine, 1-4h,6h,7h-thieno 3,2-c pyran-4-yl methanamine, 4h,6h,7h-thieno 3,2-c pyran-4-yl methanamine, 6,7-dihydro-4h-thiopheno 3,2-c pyran-4-ylmethylamine, 1-6,7-dihydro-4h-thieno 3,2-c pyran-4-yl methanamine
PubChem CID 12225896
IUPAC Name 6,7-dihydro-4H-thieno[3,2-c]pyran-4-ylmethanamine
SMILES C1COC(C2=C1SC=C2)CN

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 169.25
Percent Purity 97%
Chemical Name or Material (6, 7-Dihydro-4H-thieno[3, 2-c]pyran-4-yl)methylamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.