missing translation for 'onlineSavingsMsg'
Learn More

6,7-Dimethoxy-1,2,3,4-tetrahydroisoquinoline hydrochloride, 97%

Product Code. 11400647
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11400647 100 g 100g
11480637 5 g 5g
11490637 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11400647 Supplier Thermo Scientific Alfa Aesar Supplier No. B22469.22
 Restricted Item
Estimated Shipment: 14-10-2026
Add to basket
This item is not returnable. View return policy

CAS: 2328-12-3 | C11H16ClNO2 | 229.70 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2328-12-3
Molecular Formula C11H16ClNO2
Molecular Weight (g/mol) 229.70
MDL Number MFCD00012744
InChI Key SHOWAGCIRTUYNA-UHFFFAOYSA-N

Specifications

Melting Point 256°C to 260°C
Quantity 100 g
Beilstein 3634126
Synonym citrate iron, fe citrate
SMILES [Cl-].COC1=CC2=C(C[NH2+]CC2)C=C1OC
IUPAC Name 6,7-dimethoxy-1,2,3,4-tetrahydroisoquinolin-2-ium chloride
PubChem CID 22178220
Formula Weight 229.71
Percent Purity 97%
Chemical Name or Material 6,7-Dimethoxy-1,2,3,4-tetrahydroisoquinoline hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.